• USA Home
  • Search Results

  • 4aminotoluene3sulfonicacid187228844811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Aminotoluene-3-sulfonic acid

1 Products
Synonym:   2-Amino-5-methylbenzenesulfonic acid, 4-Methylaniline-2-sulfonic acid
Image of 4-<WBR>Aminotoluene-<WBR>3-<WBR>sulfonic acid
CAS Number:
Linear Formula:
 H2NC6H3(CH3)SO3H
Molecular Weight:
 187.22
 Beilstein/REAXYS Number:
 2211509
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More