• USA Home
  • Search Results

  • 4benzyloxybenzaldehyde21224439753911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-(Benzyloxy)benzaldehyde

1 Products
Image of 4-<WBR>(Benzyloxy)<WBR>benzaldehyde
CAS Number:
Linear Formula:
 C6H5CH2OC6H4CHO
Molecular Weight:
 212.24
 Beilstein/REAXYS Number:
 1242385
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More