• USA Home
  • Search Results

  • 4biphenylaceticacid21224572852911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Biphenylacetic acid

1 Products
Image of 4-Biphenylacetic acid
CAS Number:
Linear Formula:
 C6H5C6H4CH2CO2H
Molecular Weight:
 212.24
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More