• USA Home
  • Search Results

  • 4bromo2chlorophenol20745396456511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Bromo-2-chlorophenol

1 Products
Image of 4-<WBR>Bromo-<WBR>2-<WBR>chlorophenol
CAS Number:
Linear Formula:
 BrC6H3(Cl)OH
Molecular Weight:
 207.45
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More