• USA Home
  • Search Results

  • 4bromoanisole1870310492711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Bromoanisole

1 Products
Synonym:   1-Bromo-4-methoxybenzene
Image of 4-Bromoanisole
CAS Number:
Linear Formula:
 BrC6H4OCH3
Molecular Weight:
 187.03
 Beilstein/REAXYS Number:
 1237590
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
B56501 4-Bromoanisole, ≥99.0%,