• USA Home
  • Search Results

  • 4bromomethyl7methoxycoumarin269093523144811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Bromomethyl-7-methoxycoumarin

1 Products
Synonym:   BMC, Br-Mmc
Image of 4-<WBR>Bromomethyl-<WBR>7-<WBR>methoxycoumarin
CAS Number:
Empirical Formula (Hill Notation):
 C11H9BrO3
Molecular Weight:
 269.09
 Beilstein Registry Number:
 187211
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Fluorescence Test
Show More