• USA Home
  • Search Results

  • 4bromomethylbenzoicacid21504623288811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-(Bromomethyl)benzoic acid

1 Products
Synonym:   α-Bromo-p-toluic acid
Image of 4-<WBR>(Bromomethyl)<WBR>benzoic acid
CAS Number:
Linear Formula:
 BrCH2C6H4CO2H
Molecular Weight:
 215.04
 Beilstein/REAXYS Number:
 1862870
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More