• USA Home
  • Search Results

  • 4carvomenthenol1542556274311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Carvomenthenol

2 Products
Synonym:   4-Carvomenthenol, 4-Terpinenol
Image of 4-Carvomenthenol
CAS Number:
Empirical Formula (Hill Notation):
 C10H18O
Molecular Weight:
 154.25
 Council of Europe no.:
 2229c
 EC Number:
 FEMA Number:
 2248
 Flavis number:
 2.072
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Agency/Method
  Allergen
  Organoleptic property
  Assay
  Special Grade
Show More