• USA Home
  • Search Results

  • 4chloro2methylaniline141609569211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Chloro-2-methylaniline

2 Products
Synonym:   2-Amino-5-chlorotoluene, 4-Chloro-o-toluidine
Image of 4-<WBR>Chloro-<WBR>2-<WBR>methylaniline
CAS Number:
Linear Formula:
 ClC6H3(CH3)NH2
Molecular Weight:
 141.60
 Beilstein/REAXYS Number:
 878505
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Application(s)
Show More