• USA Home
  • Search Results

  • 4chloro2methylphenol14258157064511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Chloro-2-methylphenol

2 Products
Synonym:   4-Chloro-o-cresol
Image of 4-<WBR>Chloro-<WBR>2-<WBR>methylphenol
CAS Number:
Linear Formula:
 ClC6H3(CH3)OH
Molecular Weight:
 142.58
 Beilstein/REAXYS Number:
 1906684
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More