• USA Home
  • Search Results

  • 4chloro3nitroanisole187581029880311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Chloro-3-nitroanisole

1 Products
Synonym:   1-Chloro-4-methoxy-2-nitrobenzene
Image of 4-<WBR>Chloro-<WBR>3-<WBR>nitroanisole
CAS Number:
Linear Formula:
 ClC6H3(NO2)OCH3
Molecular Weight:
 187.58
 Beilstein/REAXYS Number:
 640872
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More