• USA Home
  • Search Results

  • 4chloro3nitrobenzoicacid201569699111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Chloro-3-nitrobenzoic acid

1 Products
Image of 4-<WBR>Chloro-<WBR>3-<WBR>nitrobenzoic acid
CAS Number:
Linear Formula:
 ClC6H3(NO2)CO2H
Molecular Weight:
 201.56
 Beilstein/REAXYS Number:
 783626
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More