• USA Home
  • Search Results

  • 4chloro3nitrophenol1735561078611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Chloro-3-nitrophenol

1 Products
Image of 4-<WBR>Chloro-<WBR>3-<WBR>nitrophenol
CAS Number:
Linear Formula:
 ClC6H3(NO2)OH
Molecular Weight:
 173.55
 Beilstein/REAXYS Number:
 2094546
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More