• USA Home
  • Search Results

  • 4chloro7nitrobenzofurazan199551019989011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Chloro-7-nitrobenzofurazan

2 Products
Synonym:   4-Chloro-7-nitro-1,2,3-benzoxadiazole, NBD-chloride
Image of 4-<WBR>Chloro-<WBR>7-<WBR>nitrobenzofurazan
CAS Number:
Empirical Formula (Hill Notation):
 C6H2ClN3O3
Molecular Weight:
 199.55
 Beilstein/REAXYS Number:
 614212
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Fluorescence Test
Show More