• USA Home
  • Search Results

  • 4chloroacetophenone154599991211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4′-Chloroacetophenone

1 Products
Image of 4′-Chloroacetophenone
CAS Number:
Linear Formula:
 ClC6H4COCH3
Molecular Weight:
 154.59
 Beilstein/REAXYS Number:
 386014
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More