• USA Home
  • Search Results

  • 4chloroaniline1275710647811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Chloroaniline

5 Products
Image of 4-Chloroaniline
CAS Number:
Linear Formula:
 ClC6H4NH2
Molecular Weight:
 127.57
 Beilstein/REAXYS Number:
 471359
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Anal. Standard Form
  Potency
  Melting Point
  Special Grade
  Assay
  Application(s)
Show More