• USA Home
  • Search Results

  • 4chlorobenzonitrile1375762303011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Chlorobenzonitrile

1 Products
Image of 4-Chlorobenzonitrile
CAS Number:
Linear Formula:
 ClC6H4CN
Molecular Weight:
 137.57
 Beilstein/REAXYS Number:
 1072122
 EC Number:
 MDL number:
 PubChem Substance ID:
 eCl@ss:
 39050680
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
115622 4-Chlorobenzonitrile, 99%,