• USA Home
  • Search Results

  • 4chlorophenylacetonitrile1515914053411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(4-Chlorophenyl)acetonitrile

1 Products
Synonym:   4-Chlorobenzyl cyanide
Image of (4-<WBR>Chlorophenyl)<WBR>acetonitrile
CAS Number:
Linear Formula:
 ClC6H4CH2CN
Molecular Weight:
 151.59
 Beilstein/REAXYS Number:
 971171
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More