• USA Home
  • Search Results

  • 4chromanol15017148193211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Chromanol

1 Products
Synonym:   3,4-Dihydro-2H-1-benzopyran-4-ol
Image of 4-Chromanol
CAS Number:
Empirical Formula (Hill Notation):
 C9H10O2
Molecular Weight:
 150.17
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
303895 4-Chromanol, ≥99%,