• USA Home
  • Search Results

  • 4dimethylaminoantipyrine231295815111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Dimethylaminoantipyrine

1 Products
Synonym:   4-Dimethylamino-2,3-dimethyl-1-phenyl-3-pyrazolin-5-one, Aminopyrine
Image of 4-<WBR>Dimethylaminoantipyrine
CAS Number:
Empirical Formula (Hill Notation):
 C13H17N3O
Molecular Weight:
 231.29
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More