• USA Home
  • Search Results

  • 4dimethylaminobenzaldehyde1491910010711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-(Dimethylamino)benzaldehyde

6 Products
Synonym:   Ehrlich’s reagent
Image of 4-<WBR>(Dimethylamino)<WBR>benzaldehyde
CAS Number:
Linear Formula:
 (CH3)2NC6H4CHO
Molecular Weight:
 149.19
 Beilstein/REAXYS Number:
 606802
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Absorption
  Vapor pressure
  Transition temperature
  Application(s)
  Potency
  Density
  Bulk Density
  Autoignition temp
  Melting Point
  Special Grade
  Solubility
Show More