• USA Home
  • Search Results

  • 4dimethylaminobenzoicacid1651961984111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-(Dimethylamino)benzoic acid

1 Products
Image of 4-<WBR>(Dimethylamino)<WBR>benzoic acid
CAS Number:
Linear Formula:
 (CH3)2NC6H4CO2H
Molecular Weight:
 165.19
 Beilstein/REAXYS Number:
 386867
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Application(s)
Show More