• USA Home
  • Search Results

  • 4dimethylaminobenzoylchloride18363475550411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-(Dimethylamino)benzoyl chloride

2 Products
Synonym:   DMABC
Image of 4-<WBR>(Dimethylamino)<WBR>benzoyl chloride
CAS Number:
Linear Formula:
 (CH3)2NC6H4COCl
Molecular Weight:
 183.63
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More