• USA Home
  • Search Results

  • 4dimethylaminopyridine12217112258311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-(Dimethylamino)pyridine

3 Products
Synonym:   N,N-Dimethylpyridin-4-amine, DMAP
Image of 4-<WBR>(Dimethylamino)<WBR>pyridine
CAS Number:
Empirical Formula (Hill Notation):
 C7H10N2
Molecular Weight:
 122.17
 Beilstein/REAXYS Number:
 110354
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Application(s)
  Purity Grade
  Potency
  Melting Point
  Solubility
  Assay
Show More