• USA Home
  • Search Results

  • 4dimethylaminopyridinepolymerbound104158294226511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-(Dimethylamino)pyridine, polymer-bound

1 Products
Image of 4-<WBR>(Dimethylamino)<WBR>pyridine, polymer-bound
CAS Number:
Empirical Formula (Hill Notation):
 C8H8
Molecular Weight:
 104.15
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Extent of labeling
Show More