• USA Home
  • Search Results

  • 4epianhydrotetracyclinehydrochloride46288446565011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Epianhydrotetracycline hydrochloride

4 Products
Synonym:   (4R,4aS,12aS)-4-(Dimethylamino)-1,4,4a,5,12,12a-hexahydro-3,10,11,12a-tetrahydroxy-6-methyl-1,12-dioxo-2-naphthacenecarboxamide monohydrochloride
Image of 4-<WBR>Epianhydrotetracycline hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C22H22N2O7 · HCl
Molecular Weight:
 462.88
 Beilstein/REAXYS Number:
 3840711
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Anal. Standard Form
  Titertest
  Physical Form
  Special Grade
Show More