• USA Home
  • Search Results

  • 4epitetracyclinehydrochloride480902331380611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Epitetracycline hydrochloride

2 Products
Synonym:   4-epi-Tetracycline hydrochloride
Image of 4-Epitetracycline hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C22H24N2O8 · HCl
Molecular Weight:
 480.90
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
  Anal. Standard Form
  Physical Form
Show More