• USA Home
  • Search Results

  • 4ethylguaiacol15219278589911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Ethylguaiacol

3 Products
Synonym:   4-Ethyl-2-methoxyphenol, NSC 82313
Image of 4-Ethylguaiacol
CAS Number:
Linear Formula:
 C2H5C6H3-2-(OCH3)OH
Molecular Weight:
 152.19
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Document Links
  Allergen
  Organoleptic property
  Titertest
  Assay
  Special Grade
Show More