• USA Home
  • Search Results

  • 4ethynylaniline117151423581511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Ethynylaniline

1 Products
Synonym:   1-Amino-4-ethynylbenzene, P-APAC
Image of 4-Ethynylaniline
CAS Number:
Linear Formula:
 HC≡CC6H4NH2
Molecular Weight:
 117.15
 Beilstein/REAXYS Number:
 2205181
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
481122 4-Ethynylaniline, 97%,