• USA Home
  • Search Results

  • 4ethynylanisole1321676860511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Ethynylanisole

1 Products
Synonym:   1-Ethynyl-4-methoxybenzene
Image of 4-Ethynylanisole
CAS Number:
Linear Formula:
 CH3OC6H4C≡CH
Molecular Weight:
 132.16
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
206490 4-Ethynylanisole, 97%,