• USA Home
  • Search Results

  • 4fluorobenzylchloride1445735211411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Fluorobenzyl chloride

1 Products
Synonym:   α-Chloro-4-fluorotoluene, 1-Chloromethyl-4-fluorobenzene
Image of 4-Fluorobenzyl chloride
CAS Number:
Linear Formula:
 FC6H4CH2Cl
Molecular Weight:
 144.57
 Beilstein/REAXYS Number:
 742272
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More