• USA Home
  • Search Results

  • 4formylbenzoicacid1501361966911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Formylbenzoic acid

1 Products
Synonym:   4-Carboxybenzaldehyde, Benzaldehyde-4-carboxylic acid, Terephthalaldehydic acid
Image of 4-Formylbenzoic acid
CAS Number:
Linear Formula:
 HO2CC6H4CHO
Molecular Weight:
 150.13
 Beilstein Registry Number:
 471734
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More

Product #
Description
124915 4-Formylbenzoic acid, 97%,