• USA Home
  • Search Results

  • 4guanidinobutyricacid1451646300311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Guanidinobutyric acid

1 Products
Synonym:   γ-Guanidinobutyric acid
Image of 4-Guanidinobutyric acid
CAS Number:
Linear Formula:
 NH=C(NH2)NHCH2CH2CH2COOH
Molecular Weight:
 145.16
 Beilstein/REAXYS Number:
 1766447
 EC Number:
 MDL number:
 NACRES:
 NA.26
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More