• USA Home
  • Search Results

  • 4hydroxy25dimethyl32hfuranone12813365877311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Hydroxy-2,5-dimethyl-3(2H)-furanone

5 Products
Synonym:   2,5-Dimethyl-4-hydroxy-3(2H)-furanone, Furaneol, Strawberry furanone
Image of 4-<WBR>Hydroxy-<WBR>2,5-<WBR>dimethyl-<WBR>3(2<I>H</I>)-furanone
CAS Number:
Empirical Formula (Hill Notation):
 C6H8O3
Molecular Weight:
 128.13
 Beilstein Registry Number:
 1281357
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
  Document Links
  Allergen
  Organoleptic property
  Biological Source
  Assay
  Special Grade
Show More