• USA Home
  • Search Results

  • 4hydroxy3methoxycinnamicacid19418113524611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Hydroxy-3-methoxycinnamic acid

1 Products
Synonym:   Ferulic acid
Image of 4-<WBR>Hydroxy-<WBR>3-<WBR>methoxycinnamic acid
CAS Number:
Empirical Formula (Hill Notation):
 C10H10O4
Molecular Weight:
 194.18
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
Show More