• USA Home
  • Search Results

  • 4hydroxy4methyl2pentanone1161612342211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Hydroxy-4-methyl-2-pentanone

3 Products
Synonym:   Diacetone alcohol
Image of 4-<WBR>Hydroxy-<WBR>4-<WBR>methyl-<WBR>2-<WBR>pentanone
CAS Number:
Linear Formula:
 (CH3)2C(OH)CH2COCH3
Molecular Weight:
 116.16
 Beilstein/REAXYS Number:
 1740440
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Application(s)
  Potency
Show More