• USA Home
  • Search Results

  • 4hydroxybenzaldehyde1221212308011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Hydroxybenzaldehyde

5 Products
Synonym:   p-Hydroxybenzaldehyde
Image of 4-<WBR>Hydroxybenzaldehyde
CAS Number:
Linear Formula:
 HOC6H4CHO
Molecular Weight:
 122.12
 Beilstein/REAXYS Number:
 471352
 EC Number:
 FEMA Number:
 3984
 Flavis number:
 5.047
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Organoleptic property
  Application(s)
  Biological Source
  Special Grade
Show More