• USA Home
  • Search Results

  • 4hydroxybenzoicacid138129996711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Hydroxybenzoic acid

7 Products
Image of 4-Hydroxybenzoic acid
CAS Number:
Linear Formula:
 HOC6H4CO2H
Molecular Weight:
 138.12
 Beilstein/REAXYS Number:
 970950
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Purity Grade
  Melting Point
  Application(s)
  Product Line
  Special Grade
  Assay
Show More