• USA Home
  • Search Results

  • 4hydroxypiperidinehydrochloride13761538217211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Hydroxypiperidine hydrochloride

1 Products
Synonym:   4-Piperidinol hydrochloride
Image of 4-Hydroxypiperidine hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C5H11NO · HCl
Molecular Weight:
 137.61
 Beilstein/REAXYS Number:
 3908995
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More