• USA Home
  • Search Results

  • 4hydroxytamoxifen387516839235811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Hydroxytamoxifen

3 Products
Synonym:   4-(1-[4-(Dimethylaminoethoxy)phenyl]-2-phenyl-1-butenyl)phenol, 4-OHT, cis/trans-4-Hydroxytamoxifen
Image of 4-Hydroxytamoxifen
CAS Number:
Empirical Formula (Hill Notation):
 C26H29NO2
Molecular Weight:
 387.51
 PubChem Substance ID:
Popular Documents:  Datasheet (PDF)  |  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Mode of Action
  Anal. Standard Form
  Activity Spectrum
  Special Grade
  Assay
  Solubility
  Physical Form
Show More