• USA Home
  • Search Results

  • 4mercaptobenzoicacid15419107436811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Mercaptobenzoic acid

2 Products
Image of 4-Mercaptobenzoic acid
CAS Number:
Linear Formula:
 HSC6H4CO2H
Molecular Weight:
 154.19
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More