• USA Home
  • Search Results

  • 4methoxy2methylaniline1371810250111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Methoxy-2-methylaniline

1 Products
Synonym:   2-Methyl-p-anisidine, m-Cresidine
Image of 4-<WBR>Methoxy-<WBR>2-<WBR>methylaniline
CAS Number:
Linear Formula:
 CH3OC6H3(CH3)NH2
Molecular Weight:
 137.18
 Beilstein/REAXYS Number:
 774727
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More