• USA Home
  • Search Results

  • 4methoxyatoluenethiol15423625860211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Methoxy-α-toluenethiol

1 Products
Synonym:   4-Methoxybenzyl mercaptan
Image of 4-Methoxy-α-toluenethiol
CAS Number:
Linear Formula:
 CH3OC6H4CH2SH
Molecular Weight:
 154.23
 Beilstein/REAXYS Number:
 471686
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More