• USA Home
  • Search Results

  • 4methoxybenzenediazoniumtetrafluoroborate2219545964311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Methoxybenzenediazonium tetrafluoroborate

1 Products
Image of 4-<WBR>Methoxybenzenediazonium tetrafluoroborate
CAS Number:
Linear Formula:
 CH3OC6H4N2BF4
Molecular Weight:
 221.95
 Beilstein/REAXYS Number:
 3579591
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More