• USA Home
  • Search Results

  • 4methoxyphenethylamine151215581211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Methoxyphenethylamine

1 Products
Synonym:   2-(4-Methoxyphenyl)ethylamine, 4-Methoxyphenethylamine
Image of 4-<WBR>Methoxyphenethylamine
CAS Number:
Linear Formula:
 CH3OC6H4CH2CH2NH2
Molecular Weight:
 151.21
 Beilstein/REAXYS Number:
 508967
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More