• USA Home
  • Search Results

  • 4methoxyphenol1241415076511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Methoxyphenol

4 Products
Synonym:   4-Hydroxyanisole, 4-MP, p-Methoxyphenol, HQMME, Hydroquinone monomethyl ether, MEHQ, MQ-F, p-Guaiacol
Image of 4-Methoxyphenol
CAS Number:
Linear Formula:
 CH3OC6H4OH
Molecular Weight:
 124.14
 Beilstein/REAXYS Number:
 507924
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  Application(s)
  Melting Point
Show More