• USA Home
  • Search Results

  • 4methyl1pentene841669137211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Methyl-1-pentene

2 Products
Image of 4-Methyl-1-pentene
CAS Number:
Linear Formula:
 (CH3)2CHCH2CH=CH2
Molecular Weight:
 84.16
 Beilstein/REAXYS Number:
 1731096
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Application(s)
Show More