• USA Home
  • Search Results

  • 4methyl2pentanone1001610810111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Methyl-2-pentanone

7 Products
Synonym:   Isobutyl methyl ketone, Isopropylacetone, MIBK, Methyl isobutyl ketone
Image of 4-Methyl-2-pentanone
CAS Number:
Linear Formula:
 (CH3)2CHCH2COCH3
Molecular Weight:
 100.16
 Beilstein/REAXYS Number:
 605399
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Refractive Index
  pH Value
  Transition temperature
  Product Line
  Ice Shipping
  Density
  Agency/Method
  Restriction
  Solubility
  Color
  UV-Absorption Values
  Vapor pressure
  Application(s)
  Special Grade
  Physical Form
Show More