• USA Home
  • Search Results

  • 4methyl3nitroaniline1521511932411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Methyl-3-nitroaniline

1 Products
Synonym:   3-Nitro-4-methylaniline, 3-Nitro-p-toluidine, 4-Amino-2-nitro-toluene
Image of 4-<WBR>Methyl-<WBR>3-<WBR>nitroaniline
CAS Number:
Linear Formula:
 CH3C6H3(NO2)NH2
Molecular Weight:
 152.15
 Beilstein/REAXYS Number:
 638984
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
Show More