• USA Home
  • Search Results

  • 4methyl5vinylthiazole12519175928011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Methyl-5-vinylthiazole

2 Products
Image of 4-<WBR>Methyl-<WBR>5-<WBR>vinylthiazole
CAS Number:
Empirical Formula (Hill Notation):
 C6H7NS
Molecular Weight:
 125.19
 Council of Europe no.:
 11633
 EC Number:
 FEMA Number:
 3313
 Flavis number:
 15.018
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Assay
Show More